(S)-Propranolol-O-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4188-1MG BICL4188-2MG
APOSBICL4188-1MGEA 447 EUR
APOSBICL4188-1MG APOSBICL4188-2MG
(S)-Propranolol-O-β-D-glucuronide ≥95%
(S)-Propranolol-O-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 58657-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR