(R)-Propranolol-O-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4189-1MG BICL4189-2MG
APOSBICL4189-1MGEA 447 EUR
APOSBICL4189-1MG APOSBICL4189-2MG
(R)-Propranolol-O-β-D-glucuronide ≥95%
(R)-Propranolol-O-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 58657-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR