(R,S)-Propranolol-O-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4190-1MG BICL4190-5MG BICL4190-2MG
APOSBICL4190-1MGEA 405 EUR
APOSBICL4190-1MG APOSBICL4190-5MG APOSBICL4190-2MG
(R,S)-Propranolol-O-β-D-glucuronide ≥95%
(R,S)-Propranolol-O-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 66322-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR