Ramipril-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4192-1MG BICL4192-2MG
APOSBICL4192-1MGEA 768 EUR
APOSBICL4192-1MG APOSBICL4192-2MG
Ramipril-acyl-β-D-glucuronide ≥95%
Ramipril-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1357570-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR