Rhein-acyl-β-D-glucuronide ≥91%

Lieferant: Apollo Scientific

BICL4194-1MG BICL4194-2MG
APOSBICL4194-1MGEA 629 EUR
APOSBICL4194-1MG APOSBICL4194-2MG
Rhein-acyl-β-D-glucuronide ≥91%
Rhein-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 190605-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR