Salicylic acid-acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxybenzoic acid-acyl-β-D-glucuronide
BICL4196-1MG
BICL4196-5MG
BICL4196-2MG
APOSBICL4196-1MGEA
393
EUR
APOSBICL4196-1MG
APOSBICL4196-5MG
APOSBICL4196-2MG
Salicylic acid-acyl-β-D-glucuronide ≥98%
Salicylic acid-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
29315-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...