Sulindac-acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific
Achtung

BICL4197-1MG BICL4197-2MG
APOSBICL4197-1MGEA 434 EUR
APOSBICL4197-1MG APOSBICL4197-2MG
Sulindac-acyl-β-D-glucuronide ≥95%
Sulindac-acyl-β-D-glucuronide
Lagertemperatur: Gefrierschrank
CAS-Nummer: 60018-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR