Sulindac sulphone-acyl-β-D-glucuronide ≥98%

Lieferant: Apollo Scientific

BICL4199-1MG BICL4199-2MG
APOSBICL4199-1MGEA 504 EUR
APOSBICL4199-1MG APOSBICL4199-2MG
Sulindac sulphone-acyl-β-D-glucuronide ≥98%
Sulindac sulphone-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 60018-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR