2,3,4,6-Tetra-O-benzoyl-β-D-glucopyranosyl fluoride ≥98%
Lieferant: Apollo Scientific
Synonyme:
2,3,4,6-Tetra-O-benzoyl-1-deoxy-1-fluoro-β-D-glucopyranoside
BICL4215-100MG
BICL4215-250MG
BICL4215-50MG
APOSBICL4215-100MGEA
447
EUR
APOSBICL4215-100MG
APOSBICL4215-250MG
APOSBICL4215-50MG
2,3,4,6-Tetra-O-benzoyl-β-D-glucopyranosyl fluoride ≥98%
2,3,4,6-Tetra-O-benzoyl-β-D-glucopyranosyl fluoride
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
4163-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...