6-O-Trityl-D-mannopyranose ≥99%

Lieferant: Apollo Scientific

Synonyme: 6-O-Triphenylmethyl-D-mannopyranose

BICL4219-1G BICL4219-500MG BICL4219-2G
APOSBICL4219-1GEA 586 EUR
APOSBICL4219-1G APOSBICL4219-500MG APOSBICL4219-2G
6-O-Trityl-D-mannopyranose ≥99%
6-O-Trityl-D-mannopyranose
Lagertemperatur: Raumtemperatur
CAS-Nummer: 160712-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR