2,3,5-Tri-O-benzyl-D-ribofuranose ≥98%

Lieferant: Apollo Scientific

Synonyme: (2R,3R,4R)-2,3,5-Tris(benzyloxy)-4-hydroxypentanal

BICL4220-5G BICL4220-1G BICL4220-2G
APOSBICL4220-5GEA 768 EUR
APOSBICL4220-5G APOSBICL4220-1G APOSBICL4220-2G
2,3,5-Tri-O-benzyl-D-ribofuranose ≥98%
2,3,5-Tri-O-benzyl-D-ribofuranose
Formel: C₂₆H₂₈O₅
Molecular Weight: 420,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425640
CAS-Nummer: 16838-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR