Zomepirac-acyl-β-D-glucuronide ≥97%
Lieferant: Apollo Scientific
BICL4221-1MG
BICL4221-5MG
BICL4221-2MG
APOSBICL4221-1MGEA
393
EUR
APOSBICL4221-1MG
APOSBICL4221-5MG
APOSBICL4221-2MG
Zomepirac-acyl-β-D-glucuronide ≥97%
Zomepirac-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
75871-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...