2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific
Achtung

BICL4226-250MG BICL4226-500MG BICL4226-100MG
APOSBICL4226-250MGEA 609 EUR
APOSBICL4226-250MG APOSBICL4226-500MG APOSBICL4226-100MG
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside ≥98%
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside
Formel: C₁₆H₂₄N₄O₉
Lagertemperatur: Gefrierschrank
MDL: MFCD19981038
CAS-Nummer: 142072-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR