Methyl 2,3,4-tri-O-acetyl-6-azido-6-deoxy-α-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

BICL4233-1G BICL4233-500MG BICL4233-2G
APOSBICL4233-1GEA 852 EUR
APOSBICL4233-1G APOSBICL4233-500MG APOSBICL4233-2G
Methyl 2,3,4-tri-O-acetyl-6-azido-6-deoxy-α-D-glucopyranoside ≥98%
Methyl 2,3,4-tri-O-acetyl-6-azido-6-deoxy-α-D-glucopyranoside
Formel: C₁₃H₁₉N₃O₈
Molecular Weight: 345.305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703846
CAS-Nummer: 21893-05-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR