4-Aminophenyl α-D-mannopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: (2R,3S,4S,5S,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

BICL4240-100MG BICL4240-250MG
APOSBICL4240-100MGEA 172 EUR
APOSBICL4240-100MG APOSBICL4240-250MG
4-Aminophenyl α-D-mannopyranoside ≥98%
4-Aminophenyl α-D-mannopyranoside
Formel: C₁₂H₁₇NO₆
Molecular Weight: 271.266 g/mol
Schmelzpunkt: 165…166 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00067366
CAS-Nummer: 34213-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR