β-D-Xylopyranosyl azide ≥98%

Lieferant: Apollo Scientific

BICL4245-250MG BICL4245-500MG BICL4245-100MG
APOSBICL4245-250MGEA 517 EUR
APOSBICL4245-250MG APOSBICL4245-500MG APOSBICL4245-100MG
β-D-Xylopyranosyl azide ≥98%
β-D-Xylopyranosyl azide
Formel: C₅H₉N₃O₄
Molecular Weight: 175,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16621028
CAS-Nummer: 51368-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR