α-D-Mannopyranosyl azide ≥95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Azido-1-deoxy-α-D-mannose
BICL4246-100MG
BICL4246-250MG
BICL4246-50MG
APOSBICL4246-100MGEA
336
EUR
APOSBICL4246-100MG
APOSBICL4246-250MG
APOSBICL4246-50MG
α-D-Mannopyranosyl azide ≥95%
α-D-Mannopyranosyl azide
Formel:
C₆H₁₁N₃O₅ Molecular Weight: 205,169 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD05865236 CAS-Nummer: 51970-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...