2,3,4-Tri-O-acetyl-β-D-xylopyranosyl azide ≥98%
Lieferant: Apollo Scientific
BICL4250-1G
BICL4250-2G
BICL4250-5G
APOSBICL4250-1GEA
393
EUR
APOSBICL4250-1G
APOSBICL4250-2G
APOSBICL4250-5G
2,3,4-Tri-O-acetyl-β-D-xylopyranosyl azide ≥98%
2,3,4-Tri-O-acetyl-β-D-xylopyranosyl azide
Formel:
C₁₁H₁₅N₃O₇ Molecular Weight: 301.253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216970 CAS-Nummer: 53784-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...