Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-α-D-glucopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme: Benzyl 4,6-O-isopropylidene-α-GlcNAc, Benzyl 2-(acetylamino)-2-deoxy-4,6-O-isopropylidene-α-D-glucopyranoside
BICL4252-1G BICL4252-250MG BICL4252-500MG
APOSBICL4252-1GEA 1270 EUR
APOSBICL4252-1G APOSBICL4252-250MG APOSBICL4252-500MG
Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-α-D-glucopyranoside ≥98%
Benzyl 2-acetamido-2-deoxy-4,6-O-isopropylidene-α-D-glucopyranoside
Formel: C₁₈H₂₅NO₆
Molecular Weight: 351.394 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269925
CAS-Nummer: 66026-10-6