Benzyl 2,3,4-tri-O-benzyl-α-D-mannopyranoside ≥98%

Lieferant: Apollo Scientific

BICL4253-250MG BICL4253-500MG BICL4253-100MG
APOSBICL4253-250MGEA 462 EUR
APOSBICL4253-250MG APOSBICL4253-500MG APOSBICL4253-100MG
Benzyl 2,3,4-tri-O-benzyl-α-D-mannopyranoside ≥98%
Benzyl 2,3,4-tri-O-benzyl-α-D-mannopyranoside
Formel: C₃₃H₃₄O₅
Molecular Weight: 510,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425507
CAS-Nummer: 57783-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR