2-Deoxy-2-fluoro-L-fucose ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dideoxy-2-fluoro-L-galactose, (2S,3R,4R,5S)-2-Fluoro-3,4,5-trihydroxyhexanal

BICL4256-10MG BICL4256-25MG BICL4256-5MG
APOSBICL4256-10MGEA 504 EUR
APOSBICL4256-10MG APOSBICL4256-25MG APOSBICL4256-5MG
2-Deoxy-2-fluoro-L-fucose ≥98%
2-Deoxy-2-fluoro-L-fucose
Formel: C₆H₁₁FO₄
Molecular Weight: 166,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457748
CAS-Nummer: 70763-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR