Propyl 2-acetamido-2-deoxy-β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: Propyl 2-(acetylamino)-2-deoxy-β-D-glucopyranoside, Propyl β-D-GlcNAc

BICL4258-100MG BICL4258-250MG BICL4258-500MG
APOSBICL4258-100MGEA 336 EUR
APOSBICL4258-100MG APOSBICL4258-250MG APOSBICL4258-500MG
Propyl 2-acetamido-2-deoxy-β-D-glucopyranoside ≥98%
Propyl 2-acetamido-2-deoxy-β-D-glucopyranoside
Formel: C₁₁H₂₁NO₆
Molecular Weight: 263,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703887
CAS-Nummer: 70832-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR