Propargyl α-D-mannopyranoside ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Propyn-1-yl α-D-mannoside, Propargyl ALPHA-D-mannopyranoside, 2-Propynyl α-D-mannopyranoside

BICL4262-100MG BICL4262-250MG BICL4262-500MG
APOSBICL4262-100MGEA 490 EUR
APOSBICL4262-100MG APOSBICL4262-250MG APOSBICL4262-500MG
Propargyl α-D-mannopyranoside ≥98%
Propargyl α-D-mannopyranoside
Formel: C₉H₁₄O₆
Molecular Weight: 218,204 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD19980990
CAS-Nummer: 854262-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR