2-Aminoethyl β-D-anthropyranoside ≥95%
Lieferant: Apollo Scientific
Synonyme:
2-Aminoethyl 4,6-dideoxy-4-[(3-hydroxy-3-methyl-1-oxobutyl)amino]-2-O-methyl-β-D-glucopyranoside
BICL4271-1MG
BICL4271-2MG
APOSBICL4271-1MGEA
1420
EUR
APOSBICL4271-1MG
APOSBICL4271-2MG
2-Aminoethyl β-D-anthropyranoside ≥95%
2-Aminoethyl β-D-anthropyranoside
Formel:
C₁₄H₂₈N₂O₆ Molecular Weight: 320,382 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122828 CAS-Nummer: 1042430-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...