2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme:
2-Azidoethyl 3,4,6-tri-O-acetyl-β-D-GalNAc, 2-Azidoethyl 3,4,6-tri-O-acetyl-2-(acetylamino)-2-deoxy-β-D-galactopyranoside
BICL4276-100MG
BICL4276-250MG
BICL4276-500MG
APOSBICL4276-100MGEA
357
EUR
APOSBICL4276-100MG
APOSBICL4276-250MG
APOSBICL4276-500MG
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside ≥98%
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside
Formel:
C₁₆H₂₄N₄O₉ Molecular Weight: 416.383 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122833 CAS-Nummer: 142072-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...