2,3,5-Tri-O-acetyl-D-arabinonic acid, γ-lactone ≥98%

Lieferant: Apollo Scientific

Synonyme: D-Arabinono-1,4-lactone triaacetate

BICL4278-1G BICL4278-2G BICL4278-5G
APOSBICL4278-1GEA 420 EUR
APOSBICL4278-1G APOSBICL4278-2G APOSBICL4278-5G
2,3,5-Tri-O-acetyl-D-arabinonic acid, γ-lactone ≥98%
2,3,5-Tri-O-acetyl-D-arabinonic acid, γ-lactone
Formel: C₁₁H₁₄O₈
Molecular Weight: 274.224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122835
CAS-Nummer: 16751-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR