1,3,4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranose ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3,4-Tri-O-acetyl-2,6-dideoxy-2-fluoro-L-galactopyranose
BICL4279-10MG
BICL4279-25MG
BICL4279-50MG
APOSBICL4279-10MGEA
281
EUR
APOSBICL4279-10MG
APOSBICL4279-25MG
APOSBICL4279-50MG
1,3,4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranose ≥98%
1,3,4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranose
Formel:
C₁₂H₁₇FO₇ Molecular Weight: 292.258 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22124251 CAS-Nummer: 188783-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...