Caffeic acid 3-O-β-D-glucopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dihydroxycinnamic acid 3-O-β-D-glucopyranoside, 3-[3-(β-D-Glucopyranosyloxy)-4-hydroxyphenyl]-2-propenoic acid
BICL4281-2MG
BICL4281-5MG
BICL4281-1MG
APOSBICL4281-2MGEA
447
EUR
APOSBICL4281-2MG
APOSBICL4281-5MG
APOSBICL4281-1MG
Caffeic acid 3-O-β-D-glucopyranoside ≥98%
Caffeic acid 3-O-β-D-glucopyranoside
Formel:
C₁₅H₁₈O₉ Molecular Weight: 342,298 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122836 CAS-Nummer: 24959-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...