Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside ≥98%

Lieferant: Apollo Scientific

BICL4283-250MG BICL4283-500MG
APOSBICL4283-250MGEA 629 EUR
APOSBICL4283-250MG APOSBICL4283-500MG
Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside ≥98%
Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside
Formel: C₇H₁₂Cl₂O₄
Molecular Weight: 231.074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457713
CAS-Nummer: 4990-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR