2,3,5-Tri-O-acetyl-D-xylonic acid, γ-lactone ≥98%
Lieferant: Apollo Scientific
Synonyme:
D-Xylono-1,4-lactone triaacetate, D-Xylonolactone triaacetate
BICL4287-1G
BICL4287-2G
BICL4287-5G
APOSBICL4287-1GEA
420
EUR
APOSBICL4287-1G
APOSBICL4287-2G
APOSBICL4287-5G
2,3,5-Tri-O-acetyl-D-xylonic acid, γ-lactone ≥98%
2,3,5-Tri-O-acetyl-D-xylonic acid, γ-lactone
Formel:
C₁₁H₁₄O₈ Molecular Weight: 274.224 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122840 CAS-Nummer: 79580-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...