Deferasirox acyl-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4292-1MG BICL4292-2MG
APOSBICL4292-1MGEA 434 EUR
APOSBICL4292-1MG APOSBICL4292-2MG
Deferasirox acyl-β-D-glucuronide ≥95%
Deferasirox acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1233196-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR