3-O-Allyl-β-D-glucopyranose ≥97%

Lieferant: Apollo Scientific

Synonyme: 3-O-2-Propen-1-yl-β-D-glucopyranose

BICL4318-10G BICL4318-25G BICL4318-5G
APOSBICL4318-10GEA 899 EUR
APOSBICL4318-10G APOSBICL4318-25G APOSBICL4318-5G
3-O-Allyl-β-D-glucopyranose ≥97%
3-O-Allyl-β-D-glucopyranose
Formel: C₉H₁₆O₆
Molecular Weight: 220,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28397791
CAS-Nummer: 77388-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR