Benzyl 3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme:
Benzyl 4,6-O-benzylidene-3-O-2-propen-1-yl-β-D-glucopyranoside
BICL4324-1G
BICL4324-500MG
BICL4324-250MG
APOSBICL4324-1GEA
991
EUR
APOSBICL4324-1G
APOSBICL4324-500MG
APOSBICL4324-250MG
Benzyl 3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98%
Benzyl 3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside
Formel:
C₂₃H₂₆O₆ Molecular Weight: 398,449 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28397794 CAS-Nummer: 103078-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...