Diosmetin 3'-O-β-D-glucuronide ≥95%
Lieferant: Apollo Scientific
Synonyme:
5-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl-D-glucopyranosiduronic acid
BICL4329-2MG
BICL4329-5MG
BICL4329-1MG
APOSBICL4329-2MGEA
671
EUR
APOSBICL4329-2MG
APOSBICL4329-5MG
APOSBICL4329-1MG
Diosmetin 3'-O-β-D-glucuronide ≥95%
Diosmetin 3'-O-β-D-glucuronide
Formel:
C₂₂H₂₀O₁₂ Molecular Weight: 476,387 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00934605 CAS-Nummer: 152503-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...