Gallic acid 3-O-β-D-glucopyranoside ≥95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(β-D-Glucopyranosyloxy)-4,5-dihydroxy-benzoic acid

BICL4337-10MG BICL4337-25MG BICL4337-5MG
APOSBICL4337-10MGEA 629 EUR
APOSBICL4337-10MG APOSBICL4337-25MG APOSBICL4337-5MG
Gallic acid 3-O-β-D-glucopyranoside ≥95%
Gallic acid 3-O-β-D-glucopyranoside
Formel: C₁₃H₁₆O₁₀
Molecular Weight: 332,26 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28397798
CAS-Nummer: 91984-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR