2,3,6,2',3',4',6'-Hepta-O-acetyl-D-maltose ≥98%

Lieferant: Apollo Scientific

Synonyme: D-Maltose heptaacetate

BICL4340-1G BICL4340-2G BICL4340-5G
APOSBICL4340-1GEA 323 EUR
APOSBICL4340-1G APOSBICL4340-2G APOSBICL4340-5G
2,3,6,2',3',4',6'-Hepta-O-acetyl-D-maltose ≥98%
2,3,6,2',3',4',6'-Hepta-O-acetyl-D-maltose
Formel: C₂₆H₃₆O₁₈
Molecular Weight: 636.553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28397800
CAS-Nummer: 56285-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR