Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: C34, TLR4-IN-C34, (2R,3S,4R,5R,6S)-5-Acetamido-2-(acetoxymethyl)-6-isopropoxytetrahydro-2H-pyran-3,4-diyl diacetate, Isopropyl 3,4,6-tri-O-acetyl-α-D-GlcNAc

BICL4343-100MG BICL4343-250MG BICL4343-50MG
APOSBICL4343-100MGEA 377 EUR
APOSBICL4343-100MG APOSBICL4343-250MG APOSBICL4343-50MG
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside ≥98%
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside
Formel: C₁₇H₂₇NO₉
Molecular Weight: 389,398 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28053519
CAS-Nummer: 40592-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR