4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxyphenyl β-D-maltopyranoside heptaacetate

BICL4357-1G BICL4357-5G BICL4357-2G
APOSBICL4357-1GEA 377 EUR
APOSBICL4357-1G APOSBICL4357-5G APOSBICL4357-2G
4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside
Formel: C₃₃H₄₂O₁₉
Molecular Weight: 742,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28397804
CAS-Nummer: 848863-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR