Methyl β-D-maltopyranoside ≥95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-O-α-D-glucopyranosyl-β-D-glucopyranoside, NSC 231821

BICL4360-1G BICL4360-2G
APOSBICL4360-1GEA 906 EUR
APOSBICL4360-1G APOSBICL4360-2G
Methyl β-D-maltopyranoside ≥95%
Methyl β-D-maltopyranoside
Formel: C₁₃H₂₄O₁₁
Molecular Weight: 356,323 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28397807
CAS-Nummer: 744-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR