Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2,3,6-tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-α-D-glucopyranosyl)-β-D-glucopyranoside
BICL4361-1G
BICL4361-2G
BICL4361-500MG
APOSBICL4361-1GEA
768
EUR
APOSBICL4361-1G
APOSBICL4361-2G
APOSBICL4361-500MG
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside
Formel:
C₂₇H₃₈O₁₈ Molecular Weight: 650,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28397808 CAS-Nummer: 13223-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...