(R,S)-Naproxen acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
Synonyme:
2-(6-Methoxynaphthalen-2-yl)propanoic acid-acyl-β-D-glucuronide
BICL4362-1MG
BICL4362-5MG
BICL4362-2MG
APOSBICL4362-1MGEA
364
EUR
APOSBICL4362-1MG
APOSBICL4362-5MG
APOSBICL4362-2MG
(R,S)-Naproxen acyl-β-D-glucuronide ≥98%
(R,S)-Naproxen acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
120143-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...