Pentyl β-D-glucopyranoside ≥98%

Lieferant: Apollo Scientific

Synonyme: Amyl β-D-glucopyranoside

BICL4363-1G BICL4363-2G BICL4363-500MG
APOSBICL4363-1GEA 796 EUR
APOSBICL4363-1G APOSBICL4363-2G APOSBICL4363-500MG
Pentyl β-D-glucopyranoside ≥98%
Pentyl β-D-glucopyranoside
Formel: C₁₁H₂₂O₆
Molecular Weight: 250.289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00057506
CAS-Nummer: 66957-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR