2,3,4,6-Tetra-O-acetyl-β-D-glucopyranose 1-salicylate ≥98%
Lieferant: Apollo Scientific
Synonyme:
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranose 1-(2-hydroxybenzoate)
BICL4367-1G
BICL4367-5G
BICL4367-2G
APOSBICL4367-1GEA
281
EUR
APOSBICL4367-1G
APOSBICL4367-5G
APOSBICL4367-2G
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranose 1-salicylate ≥98%
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranose 1-salicylate
Formel:
C₂₁H₂₄O₁₂ Molecular Weight: 468,408 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973647 CAS-Nummer: 32748-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...