3,4,6-Tri-O-acetyl-1,2-O-(1-ethoxyethylidene)-β-D-mannopyranose ≥98%

Lieferant: Apollo Scientific

Synonyme: NSC 267968, 3,4,6-Tri-O-acetyl-β-D-mannopyranose 1,2-(ethyl orthoacetate)

BICL4373-1G BICL4373-5G BICL4373-2G
APOSBICL4373-1GEA 238 EUR
APOSBICL4373-1G APOSBICL4373-5G APOSBICL4373-2G
3,4,6-Tri-O-acetyl-1,2-O-(1-ethoxyethylidene)-β-D-mannopyranose ≥98%
3,4,6-Tri-O-acetyl-1,2-O-(1-ethoxyethylidene)-β-D-mannopyranose
Formel: C₁₆H₂₄O₁₀
Molecular Weight: 376.356 g/mol
Schmelzpunkt: 103…104 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01863564
CAS-Nummer: 28140-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR