2-deoxystreptamine dihydrobromide

Lieferant: Apollo Scientific
Achtung

Synonyme: N,2-deoxystreptamine dihydrobromide

BICR109-1G BICR109-500MG BICR109-20MG
APOSBICR109-1GEA 980 EUR
APOSBICR109-1G APOSBICR109-500MG APOSBICR109-20MG
2-deoxystreptamine dihydrobromide
2-deoxystreptamine dihydrobromide
Formel: C₆H₁₄N₂O₃·2BrH
Molecular Weight: 324,011 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863375
CAS-Nummer: 84107-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR