(±)-Strigol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3aR*,5S*,8bS*,2'R*)-3-[(E)-2',5'-dihydro-4'-methyl-5'-oxo-2'-furanyloxymethylene]-5-hydroxy-8,8-dimethyl-3,3a,4,5,6,7,8,8b-octahydroindeno[1,2-b]furan-2-one

BICR133-1MG BICR133-5MG
APOSBICR133-1MGEA 545 EUR
APOSBICR133-1MG APOSBICR133-5MG
(±)-Strigol
(±)-Strigol
Formel: C₁₉H₂₂O₆
Molecular Weight: 346.374 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD27665492
CAS-Nummer: 51820-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR