2,2,2-trichloroethoxysulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N,2,2,2-trichloroethoxysulphonyl chloride, 2,2,2-trichloroethyl sulphochloridate,chlorosulphuric acid 2,2,2-trichloroethyl ester
BICR142-10G
BICR142-5G
BICR142-1G
APOSBICR142-10GEA
493
EUR
APOSBICR142-10G
APOSBICR142-5G
APOSBICR142-1G
2,2,2-trichloroethoxysulphonyl chloride
2,2,2-trichloroethoxysulphonyl chloride
Formel:
C₂H₂Cl₄O₃S Molecular Weight: 247.912 g/mol Dichte: 1.86 g/cm³ (25 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD19200066 CAS-Nummer: 764-09-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...