Quercetin-3-o-glucopyranoside
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquercitrin, Quercetin-3-O-glucoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Isoquercetin
BIDF1022-100MG
BIDF1022-250MG
APOSBIDF1022-100MGEA
49.3
EUR
APOSBIDF1022-100MG
APOSBIDF1022-250MG
Quercetin-3-o-glucopyranoside
Quercetin-3-o-glucopyranoside
Formel:
C₂₁H₂₀O₁₂ Molecular Weight: 464,376 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00017746 CAS-Nummer: 482-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...