LL-Z-1640-2

Lieferant: Apollo Scientific
Achtung

Synonyme: 5Z-7-OXOZEAENOL, JNK/p38 inhibitor

BIL0128-10MG BIL0128-5MG BIL0128-1MG
APOSBIL0128-10MGEA 596 EUR
APOSBIL0128-10MG APOSBIL0128-5MG APOSBIL0128-1MG
LL-Z-1640-2
LL-Z-1640-2
Formel: C₁₉H₂₂O₇
Molecular Weight: 362,374 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00932468
CAS-Nummer: 253863-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR