Methyl β-D-glucuronide sodium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: Sodium methyl β-D-glucopyranosiduronate

BIM0400-100MG BIM0400-1G BIM0400-500MG
APOSBIM0400-100MGEA 60.9 EUR
APOSBIM0400-100MG APOSBIM0400-1G APOSBIM0400-500MG
Methyl β-D-glucuronide sodium salt
Methyl β-D-glucuronide sodium salt
Formel: C₇H₁₁O₇·Na
Molecular Weight: 230.148 g/mol
Schmelzpunkt: 270…273 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00083405
CAS-Nummer: 134253-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR